Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCTE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283410.1 | NP_001270339.1 | 3 - 4 | 978 | 76 | |
XM_005552851.2 | XP_005552908.1 | 3 - 4 | 1462 | 76 | ||
XM_005552852.2 | XP_005552909.1 | 3 - 4 | 1745 | 76 | ||
GenBank mRNA | AB169348.1 | - | 3 - 4 | 978 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |