Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283792.1 | NP_001270721.1 | 6 - 7 | 1077 | 113 | |
GenBank mRNA | AB168617.1 | - | 6 - 7 | 1077 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |