Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283999.1 | NP_001270928.1 | 1 - 2 | 225 | 83 | |
XM_005559538.2 | XP_005559595.1 | 1 - 2 | 230 | 83 | ||
XM_005559539.2 | XP_005559596.1 | 1 - 2 | 229 | 83 | ||
XM_015452403.1 | XP_015307889.1 | 1 - 2 | 231 | 83 | ||
GenBank mRNA | AB169252.1 | - | 1 - 2 | 225 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |