Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEKT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284892.1 | NP_001271821.1 | 4 - 5 | 592 | 55 | |
XM_005543977.2 | XP_005544034.1 | 4 - 5 | 919 | 55 | ||
XM_005543978.2 | XP_005544035.1 | 4 - 5 | 564 | 55 | ||
GenBank mRNA | AB169442.1 | - | 4 - 5 | 594 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |