Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285317.1 | NP_001272246.1 | 10 - 11 | 1932 | 69 | |
XM_005544665.2 | XP_005544722.1 | 10 - 11 | 2001 | 69 | ||
GenBank mRNA | AB071053.1 | - | 1289 | 69 | ||
AB168993.1 | - | 10 - 11 | 1932 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |