Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283182.1 | NP_001270111.1 | 3 - 4 | 775 | 113 | |
XM_015440188.1 | XP_015295674.1 | 4 - 5 | 485 | 113 | ||
GenBank mRNA | AB168527.1 | - | 3 - 4 | 775 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |