Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283182.1 | NP_001270111.1 | 5 - 6 | 954 | 67 | |
XM_015440188.1 | XP_015295674.1 | 6 - 7 | 664 | 67 | ||
GenBank mRNA | AB168527.1 | - | 5 - 6 | 954 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |