Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNDC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283934.1 | NP_001270863.1 | 2 - 3 | 667 | 74 | |
GenBank mRNA | AB168957.1 | - | 2 - 3 | 667 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |