Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284680.1 | NP_001271609.1 | 2 - 3 | 260 | 96 | |
GenBank mRNA | AB169066.1 | - | 2 - 3 | 260 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |