Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQUB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283096.1 | NP_001270025.1 | 12 - 13 | 2276 | 118 | |
XM_005550638.2 | XP_005550695.1 | 13 - 14 | 2805 | 118 | ||
XM_005550639.2 | XP_005550696.1 | 11 - 12 | 2606 | 118 | ||
XM_005550640.2 | XP_005550697.1 | 11 - 12 | 2603 | 118 | ||
XM_015447303.1 | XP_015302789.1 | 12 - 13 | 2211 | 118 | ||
GenBank mRNA | AB169093.1 | - | 12 - 13 | 2276 | 118 | |
AB169129.1 | - | 10 - 11 | 1673 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |