Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEKT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285262.1 | NP_001272191.1 | 6 - 7 | 1286 | 73 | |
GenBank mRNA | AB179415.1 | - | 6 - 7 | 1286 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |