Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283284.1 | NP_001270213.1 | 3 - 4 | 262 | 115 | |
XM_015430167.1 | XP_015285653.1 | 4 - 5 | 504 | 115 | ||
GenBank mRNA | AJ426429.1 | - | 3 - 4 | 262 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |