Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283284.1 | NP_001270213.1 | 5 - 6 | 443 | 90 | |
XM_015430167.1 | XP_015285653.1 | 6 - 7 | 685 | 90 | ||
XM_015430168.1 | XP_015285654.1 | 5 - 6 | 597 | 90 | ||
GenBank mRNA | AJ426429.1 | - | 5 - 6 | 443 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Inventoried Probe spans exons |