Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THTPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283537.1 | NP_001270466.1 | 2 - 3 | 1415 | 96 | |
XM_005560890.2 | XP_005560947.1 | 2 - 3 | 1138 | 96 | ||
XM_005560891.2 | XP_005560948.1 | 2 - 3 | 1254 | 96 | ||
XM_005560893.2 | XP_005560950.1 | 2 - 3 | 751 | 96 | ||
XM_015453258.1 | XP_015308744.1 | 2 - 3 | 1616 | 96 | ||
XM_015453259.1 | XP_015308745.1 | 3 - 4 | 875 | 96 | ||
GenBank mRNA | AB055296.1 | - | 2 - 3 | 1415 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |