Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284188.1 | NP_001271117.1 | 1 | 326 | 64 | |
XM_015446618.1 | XP_015302104.1 | 2 | 802 | 64 | ||
XM_015446619.1 | XP_015302105.1 | 3 | 529 | 64 | ||
XM_015446621.1 | XP_015302107.1 | 4 | 1171 | 64 | ||
GenBank mRNA | EF514887.1 | - | 1 | 326 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |