Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285038.1 | NP_001271967.1 | 3 - 4 | 391 | 78 | |
XM_005581877.1 | XP_005581934.1 | 3 - 4 | 605 | 78 | ||
GenBank mRNA | AB083310.2 | - | 3 - 4 | 391 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |