Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2C9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287625.1 | NP_001274554.1 | 2 - 3 | 358 | 80 | |
XM_005566002.2 | XP_005566059.1 | 2 - 3 | 423 | 80 | ||
XM_005566006.2 | XP_005566063.1 | 2 - 3 | 299 | 80 | ||
GenBank mRNA | DQ074806.1 | - | 2 - 3 | 358 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Inventoried Probe spans exons |