Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CATIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287667.1 | NP_001274596.1 | 4 - 5 | 355 | 80 | |
XM_005574280.2 | XP_005574337.1 | 5 - 6 | 571 | 80 | ||
GenBank mRNA | AB071068.1 | - | 4 - 5 | 355 | 80 |