Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIBCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287709.1 | NP_001274638.1 | 2 - 3 | 646 | 87 | |
XM_015436106.1 | XP_015291592.1 | 1 - 2 | 654 | 87 | ||
GenBank mRNA | AB071099.1 | - | 2 - 3 | 646 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |