Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIBCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287709.1 | NP_001274638.1 | 6 - 7 | 1221 | 88 | |
XM_015436106.1 | XP_015291592.1 | 5 - 6 | 1229 | 88 | ||
GenBank mRNA | AB071099.1 | - | 6 - 7 | 1221 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |