Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015449302.1 | XP_015304788.1 | 12 - 13 | 3509 | 96 | |
XM_015449307.1 | XP_015304793.1 | 12 - 13 | 3509 | 96 | ||
XM_015449309.1 | XP_015304795.1 | 11 - 12 | 3367 | 96 | ||
XM_015449313.1 | XP_015304799.1 | 12 - 13 | 2776 | 96 | ||
XM_015449318.1 | XP_015304804.1 | 12 - 13 | 3515 | 96 | ||
XM_015449320.1 | XP_015304806.1 | 12 - 13 | 2777 | 96 | ||
XM_015449323.1 | XP_015304809.1 | 12 - 13 | 3510 | 96 |