Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015449302.1 | XP_015304788.1 | 36 - 37 | 7568 | 106 | |
XM_015449307.1 | XP_015304793.1 | 36 - 37 | 7568 | 106 | ||
XM_015449309.1 | XP_015304795.1 | 35 - 36 | 7426 | 106 | ||
XM_015449318.1 | XP_015304804.1 | 29 - 30 | 6623 | 106 | ||
XM_015449323.1 | XP_015304809.1 | 28 - 29 | 6210 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |