Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015442482.1 | XP_015297968.1 | 3 - 4 | 557 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |