Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SS18L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005546782.2 | XP_005546839.1 | 2 - 3 | 238 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Probe spans exons |