Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPINK8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005547010.2 | XP_005547067.1 | 3 - 4 | 493 | 85 | |
XM_015445846.1 | XP_015301332.1 | 3 - 4 | 861 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |