Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPINK8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005547010.2 | XP_005547067.1 | 4 - 5 | 551 | 101 | |
XM_015445846.1 | XP_015301332.1 | 4 - 5 | 919 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |