Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLCXD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005548191.2 | XP_005548248.1 | 3 - 4 | 1446 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |