Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005563676.2 | XP_005563733.1 | 1 - 2 | 2428 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |