Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005575072.1 | XP_005575129.1 | 6 - 7 | 642 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |