Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSC4D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015447205.1 | XP_015302691.1 | 10 - 11 | 1726 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |