Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005575731.2 | XP_005575788.1 | 5 - 6 | 2304 | 65 | |
XM_005575732.2 | XP_005575789.1 | 5 - 6 | 2294 | 65 | ||
XM_015433587.1 | XP_015289073.1 | 5 - 6 | 2030 | 65 | ||
XM_015433588.1 | XP_015289074.1 | 5 - 6 | 2036 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |