Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102122124 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005591304.2 | XP_005591361.1 | 10 - 11 | 863 | 63 | |
XM_015442330.1 | XP_015297816.1 | 10 - 11 | 912 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |