Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005541625.2 | XP_005541682.1 | 4 - 5 | 384 | 102 | |
XM_005541626.2 | XP_005541683.1 | 4 - 5 | 367 | 102 | ||
XM_005541627.2 | XP_005541684.1 | 3 - 4 | 1346 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |