Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INSL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005543121.2 | XP_005543178.1 | 1 - 2 | 248 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |