Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015431216.1 | XP_015286702.1 | 5 - 6 | 733 | 78 | |
XM_015431217.1 | XP_015286703.1 | 5 - 6 | 729 | 78 | ||
XM_015431218.1 | XP_015286704.1 | 5 - 6 | 611 | 78 | ||
XM_015431219.1 | XP_015286705.1 | 5 - 6 | 436 | 78 | ||
XM_015431220.1 | XP_015286706.1 | 5 - 6 | 734 | 78 | ||
XM_015431221.1 | XP_015286707.1 | 4 - 5 | 375 | 78 | ||
XM_015431222.1 | XP_015286708.1 | 3 - 4 | 336 | 78 | ||
XM_015431223.1 | XP_015286709.1 | 1 - 2 | 110 | 78 | ||
XM_015431228.1 | XP_015286714.1 | 1 - 2 | 176 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |