Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005594890.2 | XP_005594947.1 | 5 - 6 | 532 | 74 | |
XM_005594893.2 | XP_005594950.1 | 4 - 5 | 484 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |