Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005547526.2 | XP_005547583.1 | 2 - 3 | 149 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |