Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005565156.2 | XP_005565213.1 | 3 - 4 | 1125 | 75 | |
XM_005565157.2 | XP_005565214.1 | 2 - 3 | 1615 | 75 | ||
XM_005565158.2 | XP_005565215.1 | 2 - 3 | 1612 | 75 | ||
XM_005565159.2 | XP_005565216.1 | 2 - 3 | 1026 | 75 | ||
XM_015456049.1 | XP_015311535.1 | 4 - 5 | 1258 | 75 | ||
XM_015456050.1 | XP_015311536.1 | 3 - 4 | 1355 | 75 | ||
XM_015456052.1 | XP_015311538.1 | 3 - 4 | 1122 | 75 | ||
XM_015456053.1 | XP_015311539.1 | 2 - 3 | 1512 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |