Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SECTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005585331.2 | XP_005585388.1 | 4 - 5 | 894 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Probe spans exons |