Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015440203.1 | XP_015295689.1 | 14 - 15 | 2332 | 96 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Probe spans exons |