Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005567733.2 | XP_005567790.1 | 2 - 3 | 1540 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Probe spans exons |