Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPH14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005567809.2 | XP_005567866.1 | 6 - 7 | 988 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |