Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEPDC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015431421.1 | XP_015286907.1 | 4 - 5 | 957 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Probe spans exons |