Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015442825.1 | XP_015298311.1 | 2 | 822 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Both primers and probe map within a single exon |