Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005568389.2 | XP_005568446.1 | 3 - 4 | 1192 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |