Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005577264.2 | XP_005577321.1 | 2 - 3 | 1893 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |