Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A3GALT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005544050.2 | XP_005544107.1 | 4 - 5 | 1895 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |