Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005554304.2 | XP_005554361.1 | 3 - 4 | 1605 | 129 | |
XM_015450034.1 | XP_015305520.1 | 3 - 4 | 2091 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |