Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102123956 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015451972.1 | XP_015307458.1 | 36 - 37 | 8115 | 75 | |
XM_015451973.1 | XP_015307459.1 | 36 - 37 | 8051 | 75 | ||
XM_015451974.1 | XP_015307460.1 | 36 - 37 | 8048 | 75 | ||
XM_015451975.1 | XP_015307461.1 | 35 - 36 | 8042 | 75 | ||
XM_015451976.1 | XP_015307462.1 | 35 - 36 | 7991 | 75 | ||
XM_015451977.1 | XP_015307463.1 | 35 - 36 | 7922 | 75 | ||
XM_015451979.1 | XP_015307465.1 | 35 - 36 | 7871 | 75 | ||
XM_015451981.1 | XP_015307467.1 | 37 - 38 | 8186 | 75 | ||
XM_015451982.1 | XP_015307468.1 | 36 - 37 | 7809 | 75 | ||
XM_015451983.1 | XP_015307469.1 | 35 - 36 | 7340 | 75 | ||
XM_015451984.1 | XP_015307470.1 | 35 - 36 | 7292 | 75 | ||
XM_015451985.1 | XP_015307471.1 | 34 - 35 | 7283 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |