Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005560993.1 | XP_005561050.1 | 1 - 2 | 105 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Inventoried Probe spans exons |